Learn More
methyl 1,2,3,4-tetrahydronaphthalene-1-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 1,2,3,4-Tetrahydro-1-naphthalene-carboxylate |
| CAS | 17502-86-2 |
| Molecular Formula | C12 H14 O2 |
| Quantity | 250 mg |
| Synonym | Methyl 1,2,3,4-Tetrahydro-1-naphthalene-carboxylate |
| InChI Formula | InChI=1S/C12H14O2/c1-14-12(13)11-8-4-6-9-5-2-3-7-10(9)11/h2-3,5,7,11H,4,6,8H2,1H3 |
| SMILES | COC(=O)C1CCCc2ccccc12 |
| IUPAC Name | methyl 1,2,3,4-tetrahydronaphthalene-1-carboxylate |
| Molecular Weight (g/mol) | 190.24 |
| Formula Weight | 190.0994 |