missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate, 98%, Thermo Scientific Chemicals

Product Code. p-7030978
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467925 250 mg 250mg
15457925 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467925 Supplier Thermo Scientific Alfa Aesar Supplier No. H31888.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7355-18-2
Molecular Formula C15H20O11
Molecular Weight (g/mol) 376.314
MDL Number MFCD00069834
InChI Key DPOQCELSZBSZGX-XOBFJNJYSA-N
Synonym 2s,3r,4s,5s,6s-6-methoxycarbonyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, methyl o-tetraacetyl-beta-d-glucopyranuronate, methyl 1,2,3,4-tetra-o-acetyl-beta-d-glucuronate, methyl 1,2,3,4-tetra-o-acetyl-beta-d-glucopyranuronate, methyl 2s,3s,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxane-2-carboxylate, 1,2,3,4-tetra-o-acetyl-beta-d-glucopyranuronic acid methyl ester, methyl 1,2,3,4-tetra-o-acetyl-, a-d-glucopyranuronate, beta-d-glucopyranuronic acid, methyl ester, tetraacetate, methyl 1,2,3,4-tetra-o-acetyl-d-glucuronate, 1,2,3,4-tetra-o-acetyl-d-glucuronide methyl ester
PubChem CID 95087
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxane-2-carboxylate
SMILES CC(=O)OC1C(C(OC(C1OC(=O)C)OC(=O)C)C(=O)OC)OC(=O)C

Specifications

Melting Point 177°C to 181°C
Odor Odorless
Quantity 250 mg
Optical Rotation +8.5° (c=1 in Chloroform)
Formula Weight 376.32
Percent Purity 98%
Chemical Name or Material Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.