Learn More
Methotrexate a-tert-Butyl Ester, TRC
Product Code. 30466715
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466715 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30466715 Supplier Toronto Research Chemicals Supplier No. TRCM26075025MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methotrexate alpha-tert-Butyl Ester
CAS 79640-70-3
Molecular Formula C24 H30 N8 O5
Quantity 25 mg
Synonym N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic Acid 1-(1,1-Dimethylethyl) Ester
InChI Formula InChI=1S/C24H30N8O5/c1-24(2,3)37-22(36)16(9-10-17(33)34)29-21(35)13-5-7-15(8-6-13)32(4)12-14-11-27-20-18(28-14)19(25)30-23(26)31-20/h5-8,11,16H,9-10,12H2,1-4H3,(H,29,35)(H,33,34)(H4,25,26,27,30,31)/t16-/m0/s1
SMILES CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)OC(C)(C)C
IUPAC Name (4S)-4-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
Molecular Weight (g/mol) 510.546
Formula Weight 510.234