Learn More
Methionine-methyl-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Methionine-methyl-d3 |
| CAS | 284665-20-9 |
| Molecular Formula | C5 D3 H8 N O2 S |
| Quantity | 10 mg |
| Synonym | Methionine-methyl-d3 (9CI, ACI), L-Methionine-methyl-d3, DL-[Methyl-D3]-methionine, DL-Methionine-d3, DL-Methionine-D3 (S-methyl-D3), 2-Amino-4-(methyl-d3)sulfanylbutanoic acid, (RS)-2-Amino-4-[(methyl-d3)thio]butanoic acid, S-(Methyl-d3)-DL-homocysteine |
| InChI Formula | InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/i1D3 |
| SMILES | [2H]C([2H])([2H])SCCC(N)C(=O)O |
| IUPAC Name | 2-amino-4-(trideuteriomethylsulfanyl)butanoic acid |
| Molecular Weight (g/mol) | 152.23 |
| Formula Weight | 152.07 |