Learn More
Methionine, MedChemExpress
Product Code. 16421379
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16421379 Supplier Medchem Express Supplier No. HY13694
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Methionine (MRX-1024; D-Methionine) is an effective chemoprotective agent which can also inhibit the neuronal activity through GABAA receptor activation.
Methionine (MRX-1024
Specifications
Chemical Name or Material Methionine
Physical Form Solid
CAS 348-67-4
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C5H11NO2S
For Use With (Application) Cancer-programmed cell death
Synonym MRX-1024 D-Methionine
Solubility Information H2O : 25 mg/mL (167.55 mM; Need ultrasonic)
SMILES N[C@H](CCSC)C(O)=O
Molecular Weight (g/mol) 149.21
Formula Weight 149.21
Percent Purity 97.0%
Grade Research