missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Aminopeptidase 2/METAP2 Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90622-100UL
This item is not returnable.
View return policy
Description
Methionine Aminopeptidase 2/METAP2 Monoclonal specifically detects Methionine Aminopeptidase 2/METAP2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Methionine Aminopeptidase 2/METAP2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10988 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| eIF-2-associated p67 homolog, Initiation factor 2-associated 67 kDa glycoprotein, MAP 2, MAP2, MetAP 2, methionine aminopeptidase 2, methionyl aminopeptidase 2, MNPEP, p67, p67EC 3.4.11.18, P67EIF2, Peptidase M 2 | |
| Recombinant Mouse Methionine Aminopeptidase 2/METAP2 protein | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction