missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Aminopeptidase 1D/MAP1D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | Methionine Aminopeptidase 1D/MAP1D |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603916
|
Novus Biologicals
NBP2-99658-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632746
|
Novus Biologicals
NBP2-99658-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methionine Aminopeptidase 1D/MAP1D Polyclonal antibody specifically detects Methionine Aminopeptidase 1D/MAP1D in Human samples. It is validated for ELISASpecifications
| Methionine Aminopeptidase 1D/MAP1D | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| 0.2 um filtered solution in PBS | |
| 254042 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.11.18, MAP1DCDS of metAP-3 within PCR fragment, Metap1l, methionine aminopeptidase 1D, mitochondrial, methionyl aminopeptidase type 1D (mitochondrial), Methionyl aminopeptidase type 1D, mitochondrial | |
| Produced in rabbits immunized with purified, recombinant Human Methionine Aminopeptidase 1D/MAP1D (Accession#: NP_954697.1; Arg44-Ala335) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title