Learn More
Methasterone, TRC
Specifications
Specifications
| Chemical Name or Material | Methasterone |
| CAS | 3381-88-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21 H34 O2 |
| Quantity | 10 mg |
| Synonym | 5α-Androstan-3-one, 17β-hydroxy-2α,17-dimethyl- (6CI,7CI,8CI), 17-Methyldrostanolone, 17β-Hydroxy-2α,17-dimethyl-5α-androstan-3-one, 2α,17α-Dimethyldihydrotestosterone, Methasteron, NSC 40490, Superdrol |
| InChI Formula | InChI=1S/C21H34O2/c1-13-12-19(2)14(11-18(13)22)5-6-15-16(19)7-9-20(3)17(15)8-10-21(20,4)23/h13-17,23H,5-12H2,1-4H3/t13-,14+,15-,16+,17+,19+,20+,21+/m1/s1 |
| SMILES | C[C@@H]1C[C@@]2(C)[C@@H](CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(C)O)CC1=O |
| IUPAC Name | (2R,5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-2,10,13,17-tetramethyl-2,4,5,6,7,8,9,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 318.49 |
| Show More |