Learn More
Methanol-13C, d4, TRC
Specifications
Specifications
| Chemical Name or Material | Methanol-13C','d4 |
| CAS | 32479-98-4 |
| Molecular Formula | 13CD4O |
| Quantity | 100 mg |
| Synonym | Methan-13C-d3-ol-d |
| InChI Formula | InChI=1S/CH4O/c1-2/h2H,1H3/i1+1D3,2D |
| SMILES | [2H][13C]([2H])([2H])O[2H] |
| Molecular Weight (g/mol) | 37.06 |