Learn More
Methacrolein (Stabilized with 1% Hydroquinone), TRC
Product Code. 30133844
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133844 10 g 10g
18032550 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133844 Supplier Toronto Research Chemicals Supplier No. TRCM25880510G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78-85-3
Molecular Formula C4 H6 O
Molecular Weight (g/mol) 70.09
Synonym 2-Propenal, 2-methyl-, 2-Methyl-2-propenal (ACI), Methacrylaldehyde (8CI), 2-Formyl-1-propene, 2-Methacrolein, 2-Methyl-2-propen-1-al, 2-Methylacrolein, 2-Methylenepropanal, 2-Methylpropenal, Isobutenal, Methacrolein, Methacrylic aldehyde, Methylacrolein, Methylacrylaldehyde, NSC 8260, α-Methacrolein, α-Methylacrolein, α-Methylacrylaldehyde
IUPAC Name 2-methylprop-2-enal
SMILES CC(=C)C=O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C4H6O/c1-4(2)3-5/h3H,1H2,2H3
Formula Weight 70.0419
Chemical Name or Material Methacrolein