Learn More
Brady™ Metalized Matte Gray Polyester Labels
Product Code. 17801829
Click to view available options
Dimensions (LxW):
12.70 mm (W) x 28.70 mm (H)
25.40 mm (W) x 12.70 mm (H)
25.40 mm (W) x 50.80 mm (H)
38.10 mm (W) x 12.70 mm (H)
38.10 mm (W) x 19.05 mm (H)
Unit Size:
Pack of 140
Pack of 239
Pack of 260
Pack of 360
Pack of 480
Product Code. 17801829
Brand: Brady™ 170872
Only null left
Add to basket
Add to basket
Create tough serial and rating plate labels with the B-428 metalized polyester label material with flexible high-tack adhesive. Designed for use with durable Brady™ portable label makers.
  • Ideal for asset or panel component identification applications that require barcodes or graphics with a matte nameplate-like quality such as backer boards, DIN rail, relays, PLCs, switches and solenoids
  • Resistant to abrasions, solvents such as fuel and oil, and extreme temperatures
  • Made with permanent acrylic adhesive for excellent bonding to smooth metal and powder-coated surfaces
Specifications
Certifications/Compliance UL Recognised;CSA Accepted;AGA Approved;Halogen Free (DIN VDE 0472/part 815) Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Permanent Acrylic
Color Light Gray
Finish Matte
Format Die-cut
Material Polyester
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset tracking;Circuit board labelling;Component and equipment labelling;Data and telecommunications labelling;General identification;Inventory and inspection labelling
For Use With (Equipment) BMP51;BMP53;M511;M510
Quantity 260
Temperature Range (English) -40°F to 248°F
Temperature Range (Metric) -40°C to 120°C
Thickness (English) 0.0047 in
Thickness (Metric) 0.122 mm
Dimensions (LxW) 38.10 mm (W) x 19.05 mm (H)
Length (English) 0.75 in.
Width (English) 1.5 in
Width (Metric) 38.10 mm
Height (English) 0.75 in
Height (Metric) 19.05 mm
Alternate Part Number M5-30-428