Learn More
Lauda™ Insulating Hose
Product Code. 10001312
Change view
Click to view available options
Diameter (Metric) Inside:
10 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Unit Size
10001312 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10001312 Supplier Lauda™ Supplier No. LZM 041
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Material: Metal
  • Made of stainless steel.
  • High heat resistance
Specifications
Temperature Range -10°C to +100°C
Type Insulation Tubing
Length (Metric) 100 cm
Diameter (Metric) Inside 10 mm