Learn More
SI Analytics™ ScienceLine Metal Combination Electrode
SI Analytics™ ScienceLine Metal Combination Electrode is the first choice for titration and can be used in a range of applications like general redox titrations.
Brand: SI Analytics™ 285102002
298.55 EUR valid until 2024-05-31
Use promo code "21700" to get your promotional price.
Additional Details : Weight : 0.15000kg
Description
- Metal combination electrode with plug head and connection cable
- For use with determining acid value and halogenide (mostly chloride)
- For determinging iodine number, peroxice value, and general redox titrations
Specifications
Plug Head | |
Glass | |
Silamid™ | |
-5°C to +100°C | |
12 mm |
Platinum Diaphragm | |
Metal Combination Electrode | |
Pt 61 | |
170 mm |