Learn More
Mesitaldehyde, 97%
Product Code. p-7080539
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
Product Code. 11458830
Brand: Thermo Scientific Alfa Aesar A10104.04
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 487-68-3 | C10H12O | 148.21 g/mol
Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.
Solubility
Soluble in Chloroform
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 487-68-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00003341
InChI Key HIKRJHFHGKZKRI-UHFFFAOYSA-N
Synonym mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural
PubChem CID 10254
IUPAC Name 2,4,6-trimethylbenzaldehyde
SMILES CC1=CC(C)=C(C=O)C(C)=C1
Specifications
Melting Point 14°C
Density 1.005
Boiling Point 237°C
Flash Point 106°C (222°F)
Refractive Index 1.554
Quantity 2 g
Beilstein 1364114
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material Mesitaldehyde