Learn More
STEMCELL Technologies MesenCult™-SF Attachment Substrate
Product Code. 17108271
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 17108271
Brand: STEMCELL Technologies 05424
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Serum-free attachment substrate for human mesenchymal stem cells
  • MesenCult™-SF Attachment Substrate is a serum-free substrate for the in vitro culture of human mesenchymal stem and progenitor cells (MSCs)
  • The substrate supports both expansion of human MSCs in vitro as well as their enumeration using the colony forming unit–fibroblast (CFU-F) assay
Specifications
Cell Type Mesenchymal Stem and Progenitor Cells
Quantity 5 mg