Learn More
STEMCELL Technologies MesenCult™-SF Attachment Substrate
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
- MesenCult™-SF Attachment Substrate is a serum-free substrate for the in vitro culture of human mesenchymal stem and progenitor cells (MSCs)
- The substrate supports both expansion of human MSCs in vitro as well as their enumeration using the colony forming unit–fibroblast (CFU-F) assay
Specifications
Specifications
| Cell Type | Mesenchymal Stem and Progenitor Cells |
| Quantity | 5 mg |