Learn More
Mesaconitine, TRC
Product Code. 30135594
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135594 25 mg 25mg
30009834 10 mg 10mg
30007956 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135594 Supplier Toronto Research Chemicals Supplier No. TRCM25790025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2752-64-9
Molecular Formula C33 H45 N O11
Molecular Weight (g/mol) 631.71
SMILES COC[C@]12CN(C)C3[C@@H]4[C@H](OC)[C@H]1[C@@]3([C@H](C[C@H]2O)OC)[C@@H]5C[C@]6(O)[C@@H](OC)[C@H](O)[C@@]4(OC(=O)C)[C@H]5[C@H]6OC(=O)c7ccccc7
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C33H45NO11/c1-16(35)45-33-21-18(13-31(39,28(43-6)26(33)37)27(21)44-29(38)17-10-8-7-9-11-17)32-20(41-4)12-19(36)30(15-40-3)14-34(2)25(32)22(33)23(42-5)24(30)32/h7-11,18-28,36-37,39H,12-15H2,1-6H3/t18-,19-,20+,21-,22+,23+,24-,25?,26+,27-,28+,30+,31-,32+,33-/m1/s1
Formula Weight 631.2993
Percent Purity >95%
Chemical Name or Material Mesaconitine
Grade HPLC