Learn More
Menin-MLL inhibitor MI-2, MedChemExpress
Product Code. 16476368
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16476368
Brand: Medchem Express HY15222
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Menin-MLL inhibitor MI-2 is a Menin-MLL interaction inhibitor with IC50 of 446±28 nM.
Menin-MLL inhibitor MI-2 is a Menin-MLL interaction inhibitor with IC50 of 446±28 nM.
Specifications
Chemical Name or Material Menin-MLL inhibitor MI-2
Physical Form Solid
CAS 1271738-62-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H25N5S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (133.14 mM; Need ultrasonic)
SMILES CCCC1=CC2=C(N3CCN(C4=NCC(C)(C)S4)CC3)N=CN=C2S1
Molecular Weight (g/mol) 375.55
Formula Weight 375.55
Percent Purity 99.89%
Grade Research