Learn More
Macherey-Nagel™ Membrane Desalting Buffer MDB
Product Code. 15558471
Change view
Click to view available options
Membrane Desalting Buffer; Volume: 125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15558471 Membrane Desalting Buffer; Volume: 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15558471 Supplier Macherey-Nagel™ Supplier No. 740342.125
Please to purchase this item. Need a web account? Register with us today!
Membrane Desalting Buffer MDB to be used with NucleoSpin RNA, NucleoSpin RNA XS, NucleoSpin RNA Blood, and NucleoSpin miRNA kits
  • For NucleoSpin RNA, RNA XS, RNA Blood, and miRNA kits
Specifications
Chemical Name or Material Membrane Desaling Buffer
For Use With (Application) Isolation of RNA
For Use With (Equipment) NucleoSpin RNA, NucleoSpin RNA XS, NucleoSpin RNA Blood, and NucleoSpin miRNA Kits
Quantity 125 mL
compliance-icons
Product Identifier
  • Buffer MDB (125 mL)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: guanidium thiocyanide, ethanol