missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Mouse anti-Human, Clone: KBA.62, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14429.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human melanoma.
- High specificity
- High reactivity
Specifications
| Melanoma | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| Reacts positively against melanocytic tumors but not other tumors. | |
| Store at 4°C. | |
| IgG1 κ |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| KBA.62 | |
| Mouse monoclonal antibody raised against human melanoma. | |
| In 10 mM PBS (0.05% BSA and 0.05% azide). | |
| Human KAL cells derived from lymph node metastasis of malignant melanoma. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction