missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanocortin-5 R/MC5R Antibody (SR1772), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22481-50ul
This item is not returnable.
View return policy
Description
Melanocortin-5 R/MC5R Monoclonal antibody specifically detects Melanocortin-5 R/MC5R in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Melanocortin-5 R/MC5R | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1772 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| MC2, MC-2, MC5-R, melanocortin 5 receptor, melanocortin receptor 5, melanocortin-5 receptor | |
| A synthesized peptide derived from human Melanocortin-5 R/MC5R (Uniprot #: P33032) | |
| 50 μg | |
| GPCR | |
| 4161 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction