Learn More
MEK inhibitor, MedChemExpress
Product Code. 16497737
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16497737
Brand: Medchem Express HY12202
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress MEK inhibitor is a potent MEK inhibitor with antitumor potency.
MEK inhibitor is a potent MEK inhibitor with antitumor potency.
Specifications
Chemical Name or Material MEK inhibitor
Physical Form Solid
CAS 334951-92-7
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C26H26N4O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (117.23 mM; Need ultrasonic)
SMILES O=C(C1=CC(NC/2=O)=C(C=C1)C2=C(NC3=CC=CC(CN(C)C)=C3)/C4=CC=CC=C4)NC
Molecular Weight (g/mol) 426.51
Formula Weight 426.51
Percent Purity 98.55%
Grade Research