Learn More
STEMCELL Technologies MegaCult™-C Staining Kit for CFU-Mk
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining
- MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples
- It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods
Specifications
Specifications
| Description | Hematopoietic Stem and Progenitor Cells |
| Quantity | 1 Kit |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |