Learn More
STEMCELL Technologies MegaCult™-C Staining Kit for CFU-Mk
Product Code. 17188241
Encompass_Preferred
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 17188241
Brand: STEMCELL Technologies 04962
Please to purchase this item. Need a web account? Register with us today!
Staining kit for human CFU-Mk assays
  • MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining
  • MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples
  • It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods
Specifications
Description Hematopoietic Stem and Progenitor Cells
Quantity 1 Kit
For Use With (Application) Cell Culture, Colony Assay, Functional Assay