missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MegaCult™-C 10% BSA
Description
- MegaCult™-C Staining Kit for CFU-Mk contains all the reagents needed to stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium
- Megakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system
- The kit is also suitable for detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk)
Specifications
Specifications
| Description | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Colony Assay |
| Quantity | 6 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?