Learn More
Fisherbrand™ Mega Racks™ Tube Rack for 28 to 32 mm Tubes
Product Code. 18653983 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
18653983 Blue Each
18643983 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18653983
Only null left
Add to basket
Add to basket
High-capacity storage for your tubes
  • Fully assembled, ABS mega racks are great for high throughput labs requiring large sample storage
  • Sturdy material for easy and secure transport of sample tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack eliminating the need for multiple racks
  • Stackable for convenient refrigerated storage
  • Wells: 100
  • Well Diameter: 32 mm
  • Dimensions (L x W x H): 16.2 x 16.7 x 4.7 in. (41.2 x 42.3 x 12 cm)
Specifications
Autoclavable Not Autoclavable
Color Blue
Holds Tubes
Material ABS
Positions 100
Shape Square
Stackable Yes
Sterility Non-sterile
Dimensions (L x W x H) 16.2 x 16.7 x 4.7 in. ​(41.2 x 42.3 x 12 cm)
Product Type Tube Rack
Shelves None
Array 10 x 10