missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MeBIO, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Control analog of 6-bromoindirubin-3'-oxime ( BIO ). Displays minimal activity against CDK1/Cyclin B, GSK-3 α/β, and CDK5/p25 (IC50 values are 92.0, 44-100 and >100 μM respectively). Aryl hydrocarbon receptor (AhR) agonist; causes redistribution of AhR to the nucleus.
Active Analog also available.
Specifications
Specifications
| CAS | 667463-95-8 |
| Molecular Formula | C17H12BrN3O2 |
| Quantity | 10 mg |
| Synonym | mebio, gsk-3 inhibitor ix, control, mebio |
| InChI Key | WWVVQXIBSZPELF-JQIJEIRASA-N |
| SMILES | CN1C2=C(C=CC(=C2)Br)C(=C3C(=C4C=CC=CC4=N3)NO)C1=O |
| IUPAC Name | (3E)-6-bromo-3-[3-(hydroxyamino)indol-2-ylidene]-1-methylindol-2-one |
| Molecular Weight (g/mol) | 370.206 |
| PubChem CID | 73952600 |
| Formula Weight | 370.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction