Learn More
MDR-1339, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
MDR-1339 (DWK-1339) is an orally active and blood-brain-barrier-permeable Aβ-aggregation inhibitor, used in the research of Alzheimer's disease.
Specifications
Specifications
| Chemical Name or Material | MDR-1339 |
| CAS | 1018946-38-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H22O4 |
| Quantity | 100 mg |
| Synonym | DWK-1339 |
| Solubility Information | DMSO : 50 mg/mL (153.19 mM; Need ultrasonic) |
| SMILES | COCCCC1=CC=C(OC(C2=CC=C(OC)C(OC)=C2)=C3)C3=C1 |
| Molecular Weight (g/mol) | 326.39 |
| Show More |