Learn More
Invitrogen™ MCPT4 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142454
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
Has chymotrypsin-like activity. Hydrolyzes the amide bonds of synthetic substrates having Tyr and Phe residues at the P1 position. Preferentially hydrolyzes the 'Tyr-4-/-Ile-5' bond of angiotensin I and the 'Phe-20-/-Ala-21' bond of amyloid beta-protein, and is less active towards the 'Phe-8-/-His-9' bond of angiotensin I and the 'Phe-4-/-Ala-5' and 'Tyr-10-/-Glu-11' bonds of amyloid beta-protein. Involved in thrombin regulation and fibronectin processing.
Specifications
| MCPT4 | |
| Polyclonal | |
| Unconjugated | |
| Mcpt4 | |
| Mcpt4 | |
| Peptide with sequence C-KLQKKAKETPSVN (aa 114-126). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P21812 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 17227 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |