Learn More
MCP-4 Monoclonal Antibody (3G4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen M809
Additional Details : Weight : 0.01000kg
Description
There is one major form (long chain) and two minor forms (short chain and medium chain) of MCP-4 produced by differential signal peptide cleavage. Thus, the resulting MW of secreted MCP-4 ranges between ∽8-9kD. Western blot analysis of recombinant MCP-4/CCL13 protein detects an ∽17kD band, likely corresponding to a dimer of MCP-4. The M809 MCP-4 antibody (clone 3G4) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M810 (biotinylated conjugate of clone 8C12). The M809 MCP-4 antibody (biotinylated conjugate of clone 3G4) has also been used successfully in a sandwich ELISA as a detection antibody when paired with M810 (clone 8C12) as the coating antibody.
MCP-4 Monoclonal antibody specifically detects MCP-4 in Human samples. It is validated for ELISA, Western Blot.Specifications
MCP-4 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
Q99616 | |
CCL13 | |
Recombinant CCL13 protein | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
3G4 | |
Unconjugated | |
CCL13 | |
C-C motif chemokine 13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; C-C motif chemokine ligand 13; CCL 13; CCL13; chemokine (C-C motif) ligand 13; CKb10; CK-beta-10; H-MCP-4; MCP4; MCP-4; Monocyte chemoattractant protein 4; monocyte chemotactic protein 4; NCC1; NCC-1; new CC chemokine 1; SCYA13; SCYL1; small inducible cytokine subfamily A (Cys-Cys), member 13; small-inducible cytokine A13 | |
Mouse | |
Protein A | |
RUO | |
6357 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |