missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mCherry Polyclonal Antibody

Chicken Polyclonal Antibody
491.00 € - 1704.00 €
Specifications
| Antigen | mCherry |
|---|---|
| Concentration | Conc. Not Determined |
| Content And Storage | 4°C |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17816777
|
Invitrogen™
PA5143542 |
500 μL |
1704.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17876837
![]() |
Invitrogen™
PA5143576 |
100 μL |
491.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mCherry Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.Specifications
| mCherry | |
| 4°C | |
| Polyclonal | |
| Liquid | |
| IgY | |
| Chemical | |
| mCherry | |
| Primary |
| Conc. Not Determined | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Chicken | |
| RUO | |
| PBS with 0.02% sodium azide | |
| Full length recombinant protein expressed in and purified from E. coli. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title