missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody (235), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43768
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
mCherry Monoclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot.
Specifications
| mCherry | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| Mouse | |
| 27 kDa | |
| 0.1 mL | |
| Cellular Markers, Fusion Proteins | |
| Non-species specific | |
| Purified |
| Western Blot | |
| 235 | |
| Western Blot 1:500-1:3000 | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| This mCherry Antibody (235) was developed against full length mCherry recombinant protein. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction