Learn More
MBX-4132, MedChemExpress
Product Code. 16484648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484648 10 mg 10mg
16457367 5 mg 5mg
16455486 100 mg 100mg
16423186 25 mg 25mg
16410947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16484648 Supplier Medchem Express Supplier No. HY112565
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress MBX-4132, a member of a chemical class called oxadiazoles that inhibit trans translation by binding to the bacterial ribosome.
MBX-4132, a member of a chemical class called oxadiazoles that inhibit trans translation by binding to the bacterial ribosome.
Specifications
Chemical Name or Material MBX-4132
CAS 2286411-30-9
Color White
Purity Grade Notes Research
Molecular Formula C18H15FN4O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (73.89 mM; Need ultrasonic and warming)
SMILES O=C(NC1=NN=C(C2=CC=C(F)C=C2)O1)N3CCC4=CC=CC=C4C3
Molecular Weight (g/mol) 338.34
Formula Weight 338.34
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation