missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (rMBP/9301), Novus Biologicals™
Mouse Monoclonal Antibody
245.00 € - 486.00 €
Specifications
| Antigen | MBP |
|---|---|
| Clone | rMBP/9301 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672004
|
Novus Biologicals
NBP3-23738-20ug |
20 μg |
245.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671355
|
Novus Biologicals
NBP3-23738-100ug |
100 μg |
486.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBP Monoclonal antibody specifically detects MBP in Human, Rat, Porcine, Chicken, Guinea Pig, Mammalia, Rabbit, Sheep samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MBP | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 4155 | |
| IgG2b κ | |
| Protein A purified |
| rMBP/9301 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Pig, Chicken, Guinea Pig | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Recombinant fragment (around aa1-157) of human MBP protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title