Learn More
MBL™ Dropping Pipettes for Soda-Lime Reagent Bottles
Product Code. 15778057
Click to view available options
Description:
14/15 Joint
14/23 Joint
19/26 Joint
For Use With (Equipment):
100 mL Bottle
250 mL Bottle
50 mL Bottle
Unit Size:
Pack of 5
Product Code. 15778057
Brand: MBL SBD250
Only null left
Add to basket
Add to basket
Manufactured from clear soda-lime glass
  • With integral ground glass cone on pipette
  • Supplied with rubber teats
  • Bottles available separately
TRUSTED_SUSTAINABILITY
Specifications
Description 19/26 Joint
Material Soda Lime Glass
Includes Dropping pipettes with silicone teat
Type Pasteur Pipet
For Use With (Equipment) 250 mL Bottle