Learn More
Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kits Promotion
Shop All Thermo Scientific Products
Click to view available options
No. of Reactions:
100 reactions
20 x 20μL Reactions
Unit Size:
100 reactions
20 reactions
Description
- Increased reaction temperatures -the first strand of cDNA can be synthesized within the 42 to 65°C temperature range
- High yields of full-length first strand cDNA -with RNA templates up to 20 kb
- Flexible priming -oligo(dT)18, random hexamer, or gene-specific primers
Specifications
Specifications
| For Use With (Application) | RT-PCR |
| No. of Reactions | 100 reactions |