Learn More
Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kits Promotion
Product Code. 15777526 Shop All Thermo Scientific Products
Click to view available options
No. of Reactions:
100 reactions
20 x 20μL Reactions
Unit Size:
100 reactions
20 reactions
Product Code. 15777526
Brand: Thermo Scientific™ K1652
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Synthesize long cDNAs up to 20 kb at elevated temperatures (up to 65°C) with this complete system for highly efficient synthesis of first strand cDNA
  • Increased reaction temperatures -the first strand of cDNA can be synthesized within the 42 to 65°C temperature range
  • High yields of full-length first strand cDNA -with RNA templates up to 20 kb
  • Flexible priming -oligo(dT)18, random hexamer, or gene-specific primers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) RT-PCR
No. of Reactions 100 reactions