missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATE 2 Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26873-1-AP-150UL
This item is not returnable.
View return policy
Description
S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.Specifications
| MATE 2 | |
| Polyclonal | |
| Unconjugated | |
| SLC47A2 | |
| hMATE 2, MATE 2, MATE2, MATE2 B, MATE2 K, SLC47A2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146802, 489527 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86VL8 | |
| SLC47A2 | |
| MATE 2 Fusion Protein Ag25472 | |
| 150 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction