Learn More
Cole-Parmer™ Masterflex&trade L/S™ Platinum-Cured Silicone Pump Tubing, spooled
Reduce scrap with spooled tubing. Cole-Parmer™ Masterflex&trade L/S™ Platinum-Cured Silicone Pump Tubing features a smooth surface and low protein binding levels, ideal for pharmaceutical, biotechnology, peristaltic pumping and food applications.
Brand:  Cole-Parmer™ 96403-73
1952.25 EUR valid until 2026-06-12
BEST PRICE on Promo! Use promo code "27995" to get your promotional price.
Product Code. 11795188
  • 2055.00 € / Pack of 1
Save Big on Liquid Handling Essentials
Use promo code 27995
View all promotions
Only null left
Estimated Shipment: 24-09-2026
Add to basket
Description
Description
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
  • Ensure that you always have enough tubing on hand
  • Slightly greater clarity
  • Preferred by pharmaceutical and biotechnology industries
  • Fewer potential leachables
  • USP Class VI
  • FDA compliant