Learn More
Masterflex™ HDPE Barbed Fitting
Product Code. 12374588
Click to view available options
For Use With:
0.0625 in. Tubing I.D.
0.125 in. Tubing I.D.
0.1875 in. Tubing I.D.
0.25 in. Tubing I.D.
0.375 in. Tubing I.D.
0.5 in. Tubing I.D.
Shape:
Straight
T-shape
Y
Unit Size:
Pack of 10
Product Code. 12374588
Brand: Masterflex™ 3061401
Only null left
Add to basket
Add to basket
Ensure a tight seal every time with Masterflex fittings. Designed to tight tolerances, these fittings match perfectly to your tubing to ensure smooth flow and a proper fit.
  • Requires no tools for quick and easy installation
  • Temperature range from −53° to +65°C
  • Very good chemical resistance
  • Sterilize by ethylene oxide only
  • A variety of shapes and sizes available
  • 10/pack
Specifications
Material HDPE
For Use With 0.0625 in. Tubing I.D.
Min. Operating Temperature (Metric) -53°C
Packaging Quantity 10 Pack
Shape Y
Includes ETO sterilization
Max. Operating Temperature (Metric) 65°C
Product Type Barbed Fitting