Learn More
Thermo Scientific™ MassRuler DNA Ladders, ready-to-use
Shop All Thermo Scientific Products
Description
Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.
Highlights
- Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
Specifications
| Concentration | 6X |
| Gel Type | Agarose |
| Label or Dye | Bromophenol Blue |
| Product Type | DNA Ladder |
| Ready to Load | Yes |
| Size Range | 100 to 1000 bp |
| Gel Compatibility | Agarose Gels |
| Green Features | Sustainable packaging |
| Product Line | MassRuler |
| Quantity | 2 x 500 μL |