missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517058
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
MARK1 (MARK) is a serine/threonine kinase which is involved in cytoskeletal organization and biogenesis. Involved in cell polarity by phosphorylating the microtubule-associated proteins MAP2, MAP4 and MAPT/TAU at KXGS motifs, causing detachment from microtubules, and their disassembly. MARK1 is involved in the regulation of neuronal migration through its dual activities in regulating cellular polarity and microtubule dynamics, possibly by phosphorylating and regulating DCX. Also acts as a positive regulator of the Wnt signaling pathway, probably by mediating phosphorylation of dishevelled proteins (DVL1, DVL2 and/or DVL3).
Specifications
| MARK1 | |
| Polyclonal | |
| 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
| O08678, Q8VHJ5, Q9P0L2 | |
| MARK1 | |
| Synthetic peptide corresponding to residues surrounding Ala547 of MARK1 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| MARK1 | |
| AW491150; B930025N23Rik; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk3; KIAA1477; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity-regulating kinase 1; MARK; MARK1; mark1 {ECO:0000312; microtubule affinity regulating kinase 1; mKIAA1477; mPar-1c; PAR1 homolog c; Par1c; Par-1c; RGD:619882}; serine/threonine-protein kinase MARK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117016, 226778, 4139 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction