Learn More
Ansell Edmont™ Marigold™ Blue Natural Rubber Latex Immersion Gloves
Shop All Ansell Products
Click to view available options
Size:
7
8
Unit Size:
12 pairs
Description
- Crinkle finish provides grip in wet and dry environments
- Polyamide seamless liner
- Wrinkle natural rubber latex coating
- Gauntlet cuff
Specifications
Specifications
| Back Material | Rubber Latex |
| Certifications/Compliance | Standards EN 374, EN 407, EN 388 : 2241, 21 CFR (USA), CFIA (Canada) |
| Cuff Material | Natural Rubber Latex |
| Coating Material | Natural Rubber Latex |
| Latex Free | No |
| Product Type | Immersion Gloves |
| Size | 7 |
| Thickness (English) | 50 mil |
| Glove Cuff Style | Elasticated |
| Lining | Natural Rubber Latex |
| Show More |