missing translation for 'onlineSavingsMsg'
Learn More

MARCH3 Polyclonal Antibody

Product Code. 15778181 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15778181 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15778181 Supplier Thermo Scientific Supplier No. PA560351

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MARCH3 Polyclonal Antibody

MARCH3 Polyclonal Antibody for Western Blot, ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MARCH3
Gene Accession No. Q86UD3
Gene Alias 6330411I15Rik; A530081L18Rik; AI843639; E3 ubiquitin-protein ligase MARCH3; MARCH3; MARCH-III; membrane associated ring finger 3; membrane associated ring-CH-type finger 3; membrane-associated ring finger (C3HC4) 3; membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase; membrane-associated RING finger protein 3; Membrane-associated RING-CH protein III; RING finger protein 173; RING-type E3 ubiquitin transferase MARCH3; RNF173
Gene Symbols MARCH3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MARCH3. Recombinant protein control fragment (Product #RP-99910).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.