Learn More
Innopsys MAPIX Analyzer
Product Code. p-9224001
Click to view available options
Includes:
1-Year Single User License
3-Month Single User License
Full Single User License
Unit Size:
Each
Product Code. 16585664
Brand: Innopsys MPX.001.003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Intuitive and easy-to handle software for the quantification of microarray data from PEPperCHIP™ Peptide Microarrays
  • For microarray image analysis and data quantification
  • Intuitive and straightforward handling using .GAL files
  • Available for Windows 32 and 64 bit operating systems
Specifications
Type Software
For Use With (Application) Microarray Image Analysis and Data Quantification
For Use With (Equipment) Windows Operating Systems (32/64 Bit)
Includes 3-Month Single User License