missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K7, Mouse anti-Human, Clone: 4D2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005609-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MAASSLEQKLSRLEAKLKQENREARRRIDLNLDISPQRPRPTLQLPLANDGGSRSPSSESSPQHPTPPARPRHMLGLPSTLFTPRSMESIEIDQKLQEISpecifications
| MAP2K7 | |
| Monoclonal | |
| Unconjugated | |
| mitogen-activated protein kinase kinase 7 | |
| Jnkk2/MAPKK7/MKK7/PRKMK7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 4D2 | |
| In 1x PBS, pH 7.4 | |
| NM_145185 | |
| MAP2K7 | |
| MAP2K7 (NP_660186, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5609 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction