missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (2C4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50032SS
This item is not returnable.
View return policy
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| MAP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 1:5000 - 1:10000 | |
| P11137, P11137 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2C4 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| MAP2 Antibody (2C4) was developed against full length recombinant human MAP2D. [UniProt# P11137] | |
| 0.025 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction