missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Manyl™ Sensitive Powder-free Non-sterile Vinyl Exam Gloves

Product Code. 10351152
Click to view available options
:
Size: Large
This item is not returnable. View return policy

Product Code. 10351152

Brand: B Braun™ 6090670

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use B Braun™ Manyl™ Manyl Sensitive Powder-free Non-sterile Vinyl Exam Gloves as healthcare needs dictate to impede the spread of germs and bacteria.

  • Transparent
  • Rolled rim cuff style
  • Smooth exterior finish

Specifications

Classification PPE Category III
Color Code Transparent
Disposable Single-use
Exterior Finish Smooth
Product Type Nitrile Examination Glove
Thickness (Metric) ≥0.065 mm
Glove Cuff Style Rolled
Inspected for Medical Use Yes
Quantity 100/Pk.
Weight (Metric) .70 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.