missing translation for 'onlineSavingsMsg'
Learn More

Bode Chemie™ Manusept Basic Hand Disinfectant

Product Code. 15203049
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15203049

Brand: Bode Chemie™ 9804915

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Order Info

Quantity

Specifications

Product Type Hand Disinfectant
For Use With (Application) For hygienic and surgical rub-in hand disinfection.
Volume (Metric) 500 mL
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.