missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Manufix™ Sensitive Natural Latex Exam Gloves

Catalog Number 10742122
6.62 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.


Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Manufactured from powder-free natural rubber latex. B Braun™ Manufix™ Sensitive Natural Latex Exam Gloves are non-sterile, disposable, and suitable for use with food.

  • Manufactured from natural rubber latex
  • Single-use
  • Powder-free
  • Non-sterile
  • Suitable for use with food
Color Natural White
Disposable Single-use
Exterior Finish Micro-rough
Material Rubber Latex
Powdered Powder Free
Product Type Examination Gloves
Size Large
Thickness (Metric) ≥0.11 mm
Finger Style Textured
For Use With (Application) Suitable for food processing
Glove Cuff Style Rolled
Quantity 100 Pk.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'