Learn More
Manitimus, MedChemExpress
Product Code. 16470436
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
Product Code. 16470436
Brand: Medchem Express HY101603
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Manitimus is an inhibitor of dehydroorotate dehydrogenase, and a potent immunosuppressive drug.
Manitimus is an inhibitor of dehydroorotate dehydrogenase, and a potent immunosuppressive drug.
Specifications
Chemical Name or Material Manitimus
Physical Form Solid
CAS 202057-76-9
Color Light Yellow
Quantity 20 mg
Purity Grade Notes Research
Molecular Formula C15H11F3N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym FK778
Solubility Information DMSO : 250 mg/mL (811.00 mM; Need ultrasonic)
SMILES C#CCC/C(O)=C(C#N)/C(NC1=CC=C(C(F)(F)F)C=C1)=O.[(Z)]
Molecular Weight (g/mol) 308.26
Formula Weight 308.26
Percent Purity 97.3%
Grade Research