missing translation for 'onlineSavingsMsg'
Learn More

Manitimus, MedChemExpress

Product Code. 16466177
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16466177

Brand: Medchem Express HY101603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Manitimus is an inhibitor of dehydroorotate dehydrogenase, and a potent immunosuppressive drug.

Manitimus is an inhibitor of dehydroorotate dehydrogenase, and a potent immunosuppressive drug.

Specifications

Chemical Name or Material Manitimus
Physical Form Solid
CAS 202057-76-9
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H11F3N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym FK778
Solubility Information DMSO : 250 mg/mL (811.00 mM; Need ultrasonic)
SMILES C#CCC/C(O)=C(C#N)/C(NC1=CC=C(C(F)(F)F)C=C1)=O.[(Z)]
Molecular Weight (g/mol) 308.26
Formula Weight 308.26
Percent Purity 97.3%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.